CAS RN 133550-30-8
IUPAC Name:
N-benzyl-2-cyano-3-(3,4-dihydroxyphenyl)prop-2-enamide
Synonyms:
N-benzyl-2-cyano-3-(3,4-dihydroxyphenyl)prop-2-enamide
Molecular Weight (g/mol):
294.31
Molecular Formula:
C17H14N2O3
InChi Key:
TUCIOBMMDDOEMM-UHFFFAOYSA-N
SMILES:
OC1=C(O)C=C(C=C(C#N)C(=O)NCC2=CC=CC=C2)C=C1
1
–
3
of
3
results
1
Tocris Bioscience™ AG 490
CAS: 133550-30-8 Molecular Formula: C17H14N2O3
2
STEMCELL Technologies AG-490
Tyrosine kinase inhibitor; Inhibits EGFR, HER2, JAK2, JAK3, and STAT5a/b
3
AG490, MedChemExpress
MedChemExpress AG490 (Tyrphostin AG490) is a tyrosine kinase inhibitor that inhibits EGFR, Stat-3 and JAK2/3.