accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 132992-28-0

OHOFFF

CAS RN 132992-28-0

IUPAC Name: 2-(2,3,5-trifluorophenyl)acetic acid
Synonyms: (2,3,5-trifluorophenyl)acetic acid
Molecular Weight (g/mol): 190.12
Molecular Formula: C8H5F3O2
InChi Key: HHVKNKDKNBABCU-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC(F)=CC(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results