accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FFFHNNNH2

CAS RN 132834-58-3

IUPAC Name: 4-[5-(trifluoromethyl)pyridin-1-ium-2-yl]piperazin-1-ium
Synonyms: 4-[5-(trifluoromethyl)pyridin-1-ium-2-yl]piperazin-1-ium
Molecular Weight (g/mol): 233.24
Molecular Formula: C10H14F3N3
InChi Key: BNMSJUIMZULLAS-UHFFFAOYSA-P
SMILES: FC(F)(F)C1=CC=C([NH+]=C1)N1CC[NH2+]CC1
Boiling Point 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results