accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OH(S)OOFF

CAS RN 132741-30-1

IUPAC Name: (2S)-2-(2,4-difluorophenyl)-2-hydroxyacetate
Synonyms: (2S)-2-(2,4-difluorophenyl)-2-hydroxyacetate
Molecular Weight (g/mol): 187.12
Molecular Formula: C8H5F2O3
InChi Key: RRRQFGNNRJHLNV-ZETCQYMHSA-M
SMILES: O[C@H](C([O-])=O)C1=CC=C(F)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results