accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1327-50-0

SbTeSbTeTe

CAS RN 1327-50-0

IUPAC Name: bis(tellanylidenestibanyl) tellurium
Synonyms: antimony telluride
Molecular Weight (g/mol): 626.32
Molecular Formula: Sb2Te3
InChi Key: BPDQXJZWVBPDSN-UHFFFAOYSA-N
SMILES: [Te].[Sb]=[Te].[Sb]=[Te]
Percent Purity 
  • (4)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results