accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CHSN

CAS RN 132464-90-5

IUPAC Name: 2-(5-ethynylthiophen-2-yl)pyridine
Synonyms: 2-(5-ethynylthiophen-2-yl)pyridine
Molecular Weight (g/mol): 185.24
Molecular Formula: C11H7NS
InChi Key: CTZZEBNNWDVUFO-UHFFFAOYSA-N
SMILES: C#CC1=CC=C(S1)C1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results