accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1323966-46-6

CH3FFFFOHO

CAS RN 1323966-46-6

IUPAC Name: 2-[2-fluoro-5-methyl-4-(trifluoromethyl)phenyl]acetic acid
Synonyms: [2-fluoro-5-methyl-4-(trifluoromethyl)phenyl]acetic acid
Molecular Weight (g/mol): 236.17
Molecular Formula: C10H8F4O2
InChi Key: SCCXDMHIISSUCO-UHFFFAOYSA-N
SMILES: CC1=CC(CC(O)=O)=C(F)C=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results