accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1323966-35-3

CH3OFBrCl

CAS RN 1323966-35-3

IUPAC Name: 1-(bromomethyl)-4-chloro-2-fluoro-3-methoxybenzene
Synonyms: 1-(bromomethyl)-4-chloro-2-fluoro-3-methoxybenzene
Molecular Weight (g/mol): 253.5
Molecular Formula: C8H7BrClFO
InChi Key: PSWKNVCWOPRAMU-UHFFFAOYSA-N
SMILES: COC1=C(Cl)C=CC(CBr)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results