accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FOFFFBr

CAS RN 1323966-22-8

IUPAC Name: 1-(bromomethyl)-4-fluoro-2-(trifluoromethoxy)benzene
Synonyms: 1-(bromomethyl)-4-fluoro-2-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 273.03
Molecular Formula: C8H5BrF4O
InChi Key: YZZLXXQEOHGKHM-UHFFFAOYSA-N
SMILES: FC1=CC(OC(F)(F)F)=C(CBr)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results