accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1323966-16-0

IUPAC Name: [2-fluoro-5-methyl-4-(trifluoromethyl)phenyl]methanol
Synonyms: [2-fluoro-5-methyl-4-(trifluoromethyl)phenyl]methanol
Molecular Weight (g/mol): 208.16
Molecular Formula: C9H8F4O
InChi Key: YSXVMRQKBSXFCF-UHFFFAOYSA-N
SMILES: CC1=CC(CO)=C(F)C=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results