accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 132392-28-0

IUPAC Name: 2-bromo-1-(5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethan-1-one
Synonyms: 2-bromo-1-(5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl)ethanone
Molecular Weight (g/mol): 309.25
Molecular Formula: C16H21BrO
InChi Key: BWXPCWLVNVLIFR-UHFFFAOYSA-N
SMILES: CC1(C)CCC(C)(C)C2=C1C=CC(=C2)C(=O)CBr

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results