accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 131900-62-4

CH3ONHHN

CAS RN 131900-62-4

IUPAC Name: N-(pyrrolidin-3-yl)acetamide
Synonyms: N-(pyrrolidin-3-yl)acetamide
Molecular Weight (g/mol): 128.18
Molecular Formula: C6H12N2O
InChi Key: HDCCJUCOIKLZNM-UHFFFAOYNA-N
SMILES: CC(=O)NC1CCNC1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results