accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NOCH3NCO

CAS RN 131825-41-7

IUPAC Name: 4-isocyanato-3,5-dimethyl-1,2-oxazole
Synonyms: 4-isocyanato-3,5-dimethyl-1,2-oxazole
Molecular Weight (g/mol): 138.13
Molecular Formula: C6H6N2O2
InChi Key: JJQYAGYVYNMYGE-UHFFFAOYSA-N
SMILES: CC1=C(N=C=O)C(C)=NO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results