accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1318-93-0

H2OOOOOSiOOSiOOSiOOSiOAlAl

CAS RN 1318-93-0

IUPAC Name: dialuminium(3+) tetrakis(silanedione) hydrate trioxidandiide
Synonyms: montmorillonite
Molecular Weight (g/mol): 360.31
Molecular Formula: Al2H2O12Si4
InChi Key: GUJOJGAPFQRJSV-UHFFFAOYSA-N
SMILES: O.[O--].[O--].[O--].[Al+3].[Al+3].O=[Si]=O.O=[Si]=O.O=[Si]=O.O=[Si]=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)