accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 13173-09-6

O

CAS RN 13173-09-6

IUPAC Name: bicyclo[3.2.0]hept-2-en-6-one
Synonyms: bicyclo[3.2.0]hept-2-en-6-one
Molecular Weight (g/mol): 108.14
Molecular Formula: C7H8O
InChi Key: LNLLHUHPGPKRBM-UHFFFAOYNA-N
SMILES: O=C1CC2C=CCC12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)