CAS RN 131468-33-2
CAS RN 131468-33-2
IUPAC Name:
4,6-dichloro-N-(4-fluorophenyl)-1,3,5-triazin-2-amine
Synonyms:
4,6-dichloro-N-(4-fluorophenyl)-1,3,5-triazin-2-amine
Molecular Weight (g/mol):
259.07
Molecular Formula:
C9H5Cl2FN4
InChi Key:
GVFNRSSFMJILLU-UHFFFAOYSA-N
SMILES:
FC1=CC=C(NC2=NC(Cl)=NC(Cl)=N2)C=C1
1
–
1
of
1
results
1
2,4-Dichloro-6-(4-fluorophenylamino)-1,3,5-triazine, Thermo Scientific™
CAS: 131468-33-2 Molecular Formula: C9H5Cl2FN4 Molecular Weight (g/mol): 259.07 MDL Number: MFCD00115403 InChI Key: GVFNRSSFMJILLU-UHFFFAOYSA-N Synonym: 4,6-dichloro-n-4-fluorophenyl-1,3,5-triazin-2-amine,2,4-dichloro-6-4-fluorophenylamino-1,3,5-triazine,4,6-dichloro 1,3,5-triazin-2-yl 4-fluorophenyl amine,acmc-20amrg,1,3,5-triazin-2-amine,4,6-dichloro-n-4-fluorophenyl,2,6-dichloro-n-4-fluorophenyl-1,3,5-triazin-2-amine PubChem CID: 2779451 IUPAC Name: 4,6-dichloro-N-(4-fluorophenyl)-1,3,5-triazin-2-amine SMILES: FC1=CC=C(NC2=NC(Cl)=NC(Cl)=N2)C=C1