accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3POOCH3

CAS RN 1314246-02-0

IUPAC Name: 1-[2-(1,3-dioxolan-2-yl)phenyl]-2,5-diethylphospholane
Synonyms: 1-[2-(1,3-dioxolan-2-yl)phenyl]-2,5-diethylphospholane
Molecular Weight (g/mol): 292.36
Molecular Formula: C17H25O2P
InChi Key: UKBFKBMQNUOKTD-UHFFFAOYNA-N
SMILES: CCC1CCC(CC)P1C1=CC=CC=C1C1OCCO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results