accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1314-95-0

SnS

CAS RN 1314-95-0

IUPAC Name: stannanethione
Synonyms: tin(II) sulfide
Molecular Weight (g/mol): 152.79
Molecular Formula: H2SSn
InChi Key: SEJSGXFFJHQDAI-UHFFFAOYSA-N
SMILES: S=[SnH2]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results