accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 131076-14-7

H3COH2NNH2OCH3HClHCl

CAS RN 131076-14-7

IUPAC Name: 4,5-dimethoxybenzene-1,2-diamine dihydrochloride
Synonyms: 4,5-dimethoxybenzene-1,2-diamine dihydrochloride
Molecular Weight (g/mol): 241.11
Molecular Formula: C8H14Cl2N2O2
InChi Key: ORAAOAMEUMZGGU-UHFFFAOYSA-N
SMILES: Cl.Cl.COC1=C(OC)C=C(N)C(N)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results