CAS RN 131001-86-0
CAS RN 131001-86-0
IUPAC Name:
methyl 2-bromo-3-methylbenzoate
Synonyms:
methyl 2-bromo-3-methylbenzoate
Molecular Weight (g/mol):
229.07
Molecular Formula:
C9H9BrO2
InChi Key:
QAOFGUXVDAZKBW-UHFFFAOYSA-N
SMILES:
COC(=O)C1=C(Br)C(C)=CC=C1
1
–
1
of
1
results
1
Methyl 2-bromo-3-methylbenzoate, 98%
CAS: 131001-86-0 Molecular Formula: C9H9BrO2 Molecular Weight (g/mol): 229.07 MDL Number: MFCD00144762 InChI Key: QAOFGUXVDAZKBW-UHFFFAOYSA-N Synonym: 2-bromo-3-methyl-benzoic acid methyl ester,methyl2-bromo-3-methylbenzoate,benzoic acid, 2-bromo-3-methyl-, methyl ester,2-bromo-3-methylbenzoic acid methyl ester,methyl 2-bromo-3-methyl-benzoate,pubchem3966,acmc-1c3gk,methyl 2-bromo-m-toluate,intermediates-zcf02010,ksc495c9h PubChem CID: 10823279 IUPAC Name: methyl 2-bromo-3-methylbenzoate SMILES: COC(=O)C1=C(Br)C(C)=CC=C1