accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 13091-43-5

IUPAC Name: 2-amino-3-bromo-5-methylbenzoic acid
Synonyms: 2-amino-3-bromo-5-methylbenzoic acid
Molecular Weight (g/mol): 230.06
Molecular Formula: C8H8BrNO2
InChi Key: LCMZECCEEOQWLQ-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=C(N)C(=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results