accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 13089-11-7

IUPAC Name: methyl 3,3,3-trifluoro-2-oxopropanoate
Synonyms: methyl 3,3,3-trifluoro-2-oxopropanoate
Molecular Weight (g/mol): 156.06
Molecular Formula: C4H3F3O3
InChi Key: XGLLQDIWQRQROJ-UHFFFAOYSA-N
SMILES: COC(=O)C(=O)C(F)(F)F
Boiling Point 
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)