accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CoOCoOOCoO

CAS RN 1308-06-1

IUPAC Name: oxo[(oxocobaltio)oxy]cobalt; oxocobalt
Synonyms: Cobalt(II,III) oxide
Molecular Weight (g/mol): 240.8
Molecular Formula: Co3O4
InChi Key: LBFUKZWYPLNNJC-UHFFFAOYSA-N
SMILES: O=[Co].O=[Co]O[Co]=O
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
Physical Form 
  • (12)
Percent Purity 
  • (3)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (15)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (12)
  • (3)
  • (2)
  • (2)