accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OONH2

CAS RN 13066-01-8

IUPAC Name: 4-(2-methoxyphenoxy)aniline
Synonyms: 4-(2-methoxyphenoxy)aniline
Molecular Weight (g/mol): 215.25
Molecular Formula: C13H13NO2
InChi Key: XFOFRBMGVDBINH-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1OC1=CC=C(N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results