accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1306-05-4

OPOOOOPOOOOPOOOFCaCaCaCaCa

CAS RN 1306-05-4

IUPAC Name: pentacalcium fluoride triphosphate
Synonyms: pentacalcium fluoride triphosphate
Molecular Weight (g/mol): 504.3
Molecular Formula: Ca5FO12P3
InChi Key: VSIIXMUUUJUKCM-UHFFFAOYSA-D
SMILES: [F-].[Ca++].[Ca++].[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results