accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 130309-37-4

NOOOHO

CAS RN 130309-37-4

IUPAC Name: 1-{[(9H-fluoren-9-yl)methoxy]carbonyl}-octahydro-1H-indole-2-carboxylic acid
Synonyms: 1-[(9H-fluoren-9-ylmethoxy)carbonyl]-octahydroindole-2-carboxylic acid
Molecular Weight (g/mol): 391.47
Molecular Formula: C24H25NO4
InChi Key: JBZXLQHJZHITMW-UHFFFAOYNA-N
SMILES: OC(=O)C1CC2CCCCC2N1C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results