accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1303-58-8

OOOAuAu

CAS RN 1303-58-8

IUPAC Name: digold(3+) trioxidandiide
Synonyms: gold(III) oxide
Molecular Weight (g/mol): 441.93
Molecular Formula: Au2O3
InChi Key: DDYSHSNGZNCTKB-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[Au+3].[Au+3]
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results