accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1303-28-2

OAsOOAsOO

CAS RN 1303-28-2

IUPAC Name: diarsooxidane
Synonyms: arsenic pentoxide diarsenic pentaoxide
Molecular Weight (g/mol): 229.84
Molecular Formula: As2O5
InChi Key: COHDHYZHOPQOFD-UHFFFAOYSA-N
SMILES: O=[As](=O)O[As](=O)=O
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)