accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1303-11-3

AsIn

CAS RN 1303-11-3

IUPAC Name: indiganylidynearsane
Synonyms: indium arsenide
Molecular Weight (g/mol): 189.74
Molecular Formula: AsIn
InChi Key: RPQDHPTXJYYUPQ-UHFFFAOYSA-N
SMILES: [As]#[In]
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results