accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 13026-12-5

IUPAC Name: (2E)-3-(naphthalen-1-yl)prop-2-enoic acid
Synonyms: (2E)-3-(naphthalen-1-yl)prop-2-enoic acid
Molecular Weight (g/mol): 198.22
Molecular Formula: C13H10O2
InChi Key: WPXMLUUYWNHQOR-CMDGGOBGSA-N
SMILES: OC(=O)\C=C\C1=C2C=CC=CC2=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results