accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1302-82-5

AlAlSeSeSe

CAS RN 1302-82-5

IUPAC Name: dialuminium(3+) triselandiide
Synonyms: aluminium selenide
Molecular Weight (g/mol): 290.88
Molecular Formula: Al2Se3
InChi Key: CYRGZAAAWQRSMF-UHFFFAOYSA-N
SMILES: [Al+3].[Al+3].[Se--].[Se--].[Se--]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results