accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1301739-00-3

H3CONNNH2HCl

CAS RN 1301739-00-3

IUPAC Name: 1-(3-methoxypyridin-2-yl)piperidin-4-amine hydrochloride
Synonyms: 1-(3-methoxypyridin-2-yl)piperidin-4-amine hydrochloride
Molecular Weight (g/mol): 243.74
Molecular Formula: C11H18ClN3O
InChi Key: QBVBNVDHWTXFAT-UHFFFAOYSA-N
SMILES: Cl.COC1=C(N=CC=C1)N1CCC(N)CC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results