accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 129842-38-2

IUPAC Name: 1-chloro-4-(3,4-dimethylphenyl)phthalazine
Synonyms: 1-chloro-4-(3,4-dimethylphenyl)phthalazine
Molecular Weight (g/mol): 268.74
Molecular Formula: C16H13ClN2
InChi Key: GYYXXMBDNZTEND-UHFFFAOYSA-N
SMILES: CC1=C(C)C=C(C=C1)C1=C2C=CC=CC2=C(Cl)N=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results