CAS RN 129414-26-2
CAS RN 129414-26-2
IUPAC Name:
methyl 4-(pyrrolidin-1-yl)benzoate
Synonyms:
methyl 4-(pyrrolidin-1-yl)benzoate
Molecular Weight (g/mol):
205.26
Molecular Formula:
C12H15NO2
InChi Key:
XNIVNLQKURGIAY-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC=C(C=C1)N1CCCC1