accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 129-44-2

NH2ONH2O

CAS RN 129-44-2

IUPAC Name: 1,5-diamino-9,10-dihydroanthracene-9,10-dione
Synonyms: 1,5-diaminoanthracene-9,10-dione
Molecular Weight (g/mol): 238.25
Molecular Formula: C14H10N2O2
InChi Key: VWBVCOPVKXNMMZ-UHFFFAOYSA-N
SMILES: NC1=CC=CC2=C1C(=O)C1=CC=CC(N)=C1C2=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results