accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

SSOOCl

CAS RN 128852-17-5

IUPAC Name: 4,5,6,7-tetrahydro-1-benzothiophene-2-sulfonyl chloride
Synonyms: 4,5,6,7-tetrahydro-1-benzothiophene-2-sulfonyl chloride
Molecular Weight (g/mol): 236.73
Molecular Formula: C8H9ClO2S2
InChi Key: UUBSHLWNADKCBU-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC2=C(CCCC2)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results