accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1282805-05-3

CH3OONHBr

CAS RN 1282805-05-3

IUPAC Name: N-[(4-bromophenyl)methyl]-2-methoxybenzamide
Synonyms: N-[(4-bromophenyl)methyl]-2-methoxybenzamide
Molecular Weight (g/mol): 320.19
Molecular Formula: C15H14BrNO2
InChi Key: YLBPDHZRDAOTDN-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1C(=O)NCC1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results