accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 127729-35-5

CH3POHOH3N

CAS RN 127729-35-5

IUPAC Name: (3-azaniumylpropyl)(methyl)phosphinic acid
Synonyms: 3-ammoniopropyl(methyl)phosphinic acid
Molecular Weight (g/mol): 138.13
Molecular Formula: C4H13NO2P
InChi Key: NHVRIDDXGZPJTJ-UHFFFAOYNA-O
SMILES: CP(O)(=O)CCC[NH3+]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results