accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 126956-48-7

OOOONi

CAS RN 126956-48-7

IUPAC Name: nickel(2+) hydrate oxalate
Synonyms: nickel(2+) hydrate oxalate
Molecular Weight (g/mol): 164.73
Molecular Formula: C2H2NiO5
InChi Key: DCEDPLWSSQQNGQ-UHFFFAOYSA-L
SMILES: O.[Ni++].[O-]C(=O)C([O-])=O
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)