accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FClOFFF

CAS RN 126917-10-0

IUPAC Name: 2-fluoro-4-(trifluoromethyl)benzoyl chloride
Synonyms: 2-fluoro-4-(trifluoromethyl)benzoyl chloride
Molecular Weight (g/mol): 226.55
Molecular Formula: C8H3ClF4O
InChi Key: OOAHPLWBUUTFMV-UHFFFAOYSA-N
SMILES: FC1=CC(=CC=C1C(Cl)=O)C(F)(F)F
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)