accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

BNHH3CFFF

CAS RN 1268341-00-9

IUPAC Name: {[benzyl(methyl)azaniumyl]methyl}trifluoroboranuide
Synonyms: {[benzyl(methyl)ammonio]methyl}trifluoroboranuide
Molecular Weight (g/mol): 203.02
Molecular Formula: C9H13BF3N
InChi Key: JRTRNQRZOUGDCI-UHFFFAOYNA-O
SMILES: C[NH+](CC1=CC=CC=C1)C[B-](F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results