accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3I

CAS RN 126261-84-5

IUPAC Name: 1-iodo-4-propylbenzene
Synonyms: 1-iodo-4-propylbenzene
Molecular Weight (g/mol): 246.09
Molecular Formula: C9H11I
InChi Key: ZRWCHMFGMKNWEC-UHFFFAOYSA-N
SMILES: CCCC1=CC=C(I)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results