accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1261828-46-9

IUPAC Name: 2-methyl-5-(trifluoromethoxy)benzamide
Synonyms: 2-methyl-5-(trifluoromethoxy)benzamide
Molecular Weight (g/mol): 219.16
Molecular Formula: C9H8F3NO2
InChi Key: JHVXNWZOAYHTHU-UHFFFAOYSA-N
SMILES: CC1=C(C=C(OC(F)(F)F)C=C1)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results