accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1261653-03-5

H3CONOFFF

CAS RN 1261653-03-5

IUPAC Name: 4-methoxy-3-(trifluoromethoxy)benzonitrile
Synonyms: 4-methoxy-3-(trifluoromethoxy)benzonitrile
Molecular Weight (g/mol): 217.15
Molecular Formula: C9H6F3NO2
InChi Key: DDKKATZYULLWQH-UHFFFAOYSA-N
SMILES: COC1=C(OC(F)(F)F)C=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results