CAS RN 1261647-71-5
CAS RN 1261647-71-5
IUPAC Name:
1-[3-methoxy-2-(trifluoromethyl)phenyl]methanamine
Synonyms:
1-[3-methoxy-2-(trifluoromethyl)phenyl]methanamine
Molecular Weight (g/mol):
205.18
Molecular Formula:
C9H10F3NO
InChi Key:
FIQKCMDETAERAO-UHFFFAOYSA-N
SMILES:
COC1=CC=CC(CN)=C1C(F)(F)F
1
–
1
of
1
results
1
3-Methoxy-2-(trifluoromethyl)benzylamine, 97%, Thermo Scientific™
CAS: 1261647-71-5 Molecular Formula: C9H10F3NO Molecular Weight (g/mol): 205.18 MDL Number: MFCD18398592 InChI Key: FIQKCMDETAERAO-UHFFFAOYSA-N Synonym: 3-methoxy-2-trifluoromethyl phenyl methanamine,3-methoxy-2-trifluoromethyl benzylamine,1-3-methoxy-2-trifluoromethyl phenyl methanamine PubChem CID: 73012217 IUPAC Name: [3-methoxy-2-(trifluoromethyl)phenyl]methanamine SMILES: COC1=CC=CC(CN)=C1C(F)(F)F