accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OFFFOCH3

CAS RN 1261444-86-3

IUPAC Name: 2-methoxy-1-methyl-4-(trifluoromethoxy)benzene
Synonyms: 2-methoxy-1-methyl-4-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 206.16
Molecular Formula: C9H9F3O2
InChi Key: XYFKOVVUHAFOSZ-UHFFFAOYSA-N
SMILES: COC1=C(C)C=CC(OC(F)(F)F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results