accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1256483-51-8

SNH2NHNSNH2

CAS RN 1256483-51-8

IUPAC Name: 2-(3-carbamothioylphenyl)-1H-1,3-benzodiazole-6-carbothioamide
Synonyms: 2-(3-carbamothioylphenyl)-3H-1,3-benzodiazole-5-carbothioamide
Molecular Weight (g/mol): 312.41
Molecular Formula: C15H12N4S2
InChi Key: XKUFJCIMNRMHRY-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC=C2N=C(NC2=C1)C1=CC(=CC=C1)C(N)=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results