accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

BOOH3CCH3H3CH3CHNOOCH3CH3CH3F

CAS RN 1256256-45-7

IUPAC Name: tert-butyl N-[3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate
Synonyms: tert-butyl N-[3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate
Molecular Weight (g/mol): 337.2
Molecular Formula: C17H25BFNO4
InChi Key: FALNYBWTRBHWID-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1=CC(F)=C(C=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results