accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 125464-42-8

NH2SOHOOCl

CAS RN 125464-42-8

IUPAC Name: 3-amino-2-(4-chlorophenyl)propane-1-sulfonic acid
Synonyms: saclofen
Molecular Weight (g/mol): 249.71
Molecular Formula: C9H12ClNO3S
InChi Key: JYLNVJYYQQXNEK-UHFFFAOYNA-N
SMILES: NCC(CS(O)(=O)=O)C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results