CAS RN 124900-72-7
CAS RN 124900-72-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
YM17E, MedChemExpress
MedChemExpress YM17E is an inhibitor of acyl CoA:cholesterol acyltransferase (ACAT), with IC50 of 44 nM in rabbit liver microsomes in vitro.